missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxynaphthalene-2-boronic acid, 95%

Catalog No. AAL1906006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1906006 5 g
AAL1906003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1906006 Supplier Thermo Scientific Chemicals Supplier No. L1906006

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 156641-98-4 | C11H11BO3 | 202.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 156641-98-4
Molecular Formula C11H11BO3
Molecular Weight (g/mol) 202.02
MDL Number MFCD03093087
InChI Key GZFAVYWCPSMLCM-UHFFFAOYSA-N
Synonym 6-methoxy-2-naphthaleneboronic acid, 6-methoxynaphthalen-2-yl boronic acid, 6-methoxy-2-naphthylboronic acid, 6-methoxynaphthalene-2-boronic acid, 6-methoxy-2-naphthyl boronic acid, 2-methoxynaphthalene-6-boronic acid, boronic acid, 6-methoxy-2-naphthalenyl, 6-methoxy-2-naphthalenyl boronic acid, acmc-209deh, ksc489k9r
PubChem CID 4641692
IUPAC Name (6-methoxynaphthalen-2-yl)boronic acid
SMILES COC1=CC2=CC=C(C=C2C=C1)B(O)O

Specifications

Melting Point ∽245°C (decomposition)
Quantity 5 g
Beilstein 7636998
Formula Weight 202.02
Percent Purity 95%
Chemical Name or Material 6-Methoxynaphthalene-2-boronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only