missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxy-2-naphthaleneboronic acid, 97%

Catalog No. AC399860050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC399860050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC399860050 Supplier Thermo Scientific Chemicals Supplier No. 399860050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 156641-98-4 | C11H11BO3 | 202.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 156641-98-4
Molecular Formula C11H11BO3
Molecular Weight (g/mol) 202.02
MDL Number MFCD03093087
InChI Key GZFAVYWCPSMLCM-UHFFFAOYSA-N
Synonym 6-methoxy-2-naphthaleneboronic acid, 6-methoxynaphthalen-2-yl boronic acid, 6-methoxy-2-naphthylboronic acid, 6-methoxynaphthalene-2-boronic acid, 6-methoxy-2-naphthyl boronic acid, 2-methoxynaphthalene-6-boronic acid, boronic acid, 6-methoxy-2-naphthalenyl, 6-methoxy-2-naphthalenyl boronic acid, acmc-209deh, ksc489k9r
PubChem CID 4641692
IUPAC Name (6-methoxynaphthalen-2-yl)boronic acid
SMILES COC1=CC2=CC=C(C=C2C=C1)B(O)O

Specifications

Melting Point >295°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 5 g
Solubility Information Solubility in water: ca 2.5%
Formula Weight 202.04
Percent Purity 97%
Chemical Name or Material 6-Methoxy-2-naphthaleneboronic acid
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only