accessibility menu, dialog, popup

UserName

CAS RN 15658-35-2

IUPAC Name: 6-[(5-carboxypyridin-2-yl)disulfanyl]pyridine-3-carboxylic acid
Synonyms: 6,6'-dithiodinicotinic acid
Molecular Weight (g/mol): 308.33
Molecular Formula: C12H8N2O4S2
InChi Key: GSASOFRDSIKDSN-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(SSC2=CC=C(C=N2)C(O)=O)N=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results