CAS RN 15599-91-4
CAS RN 15599-91-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Hexafluorocyclotriphosphazene 97.0+%, TCI America™
CAS: 15599-91-4 Molecular Formula: F6N3P3 Molecular Weight (g/mol): 248.93 MDL Number: MFCD00192400 InChI Key: DKQPXAWBVGCNHG-UHFFFAOYSA-N PubChem CID: 139958 IUPAC Name: hexafluoro-1,3,5,2λ⁵,4λ⁵,6λ⁵-triazatriphosphinine SMILES: FP1(F)=NP(F)(F)=NP(F)(F)=N1