accessibility menu, dialog, popup

UserName

CAS RN 1556-18-9

IUPAC Name: iodocyclopentane
Synonyms: cyclopentane, iodo-
Molecular Weight (g/mol): 196.03
Molecular Formula: C5H9I
InChi Key: PCEBAZIVZVIQEO-UHFFFAOYSA-N
SMILES: IC1CCCC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)