CAS RN 1552-92-7
IUPAC Name:
ethyl 2-cyclohexylideneacetate
Synonyms:
ethyl 2-cyclohexylideneacetate
Molecular Weight (g/mol):
168.24
Molecular Formula:
C10H16O2
InChi Key:
MCWDXHYYYNGYGK-UHFFFAOYSA-N
SMILES:
CCOC(=O)C=C1CCCCC1
1
–
1
of
1
results
1
Promotions Available
Ethyl cyclohexylideneacetate, 98%
Ethyl cyclohexylideneacetate, 98%
CAS: 1552-92-7 Molecular Formula: C10H16O2 Molecular Weight (g/mol): 168.24 MDL Number: MFCD00017627,MFCD00963857 InChI Key: MCWDXHYYYNGYGK-UHFFFAOYSA-N Synonym: ethyl cyclohexylideneacetate,acetic acid, cyclohexylidene-, ethyl ester,ethyl 2-cyclohexylidenacetate,cyclohexylideneacetic acid ethyl ester,ethoxycarbonylmethylenecyclohexane,.delta.1,.alpha.-cyclohexaneacetic acid, ethyl ester,acmc-20ahxr,ethylcyclohexylidenacetate,.delta.1, ethyl ester,ethyl2-cyclohexylideneacetate PubChem CID: 73776 IUPAC Name: ethyl 2-cyclohexylideneacetate SMILES: CCOC(=O)C=C1CCCCC1