accessibility menu, dialog, popup

UserName

H2NOFFNH2FF

CAS RN 1548-74-9

IUPAC Name: 4-amino-2,3,5,6-tetrafluorobenzamide
Synonyms: 4-amino-2,3,5,6-tetrafluorobenzamide
Molecular Weight (g/mol): 208.12
Molecular Formula: C7H4F4N2O
InChi Key: CAERPAFTLPIDJT-UHFFFAOYSA-N
SMILES: NC(=O)C1=C(F)C(F)=C(N)C(F)=C1F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results