accessibility menu, dialog, popup

UserName

H3COOHNNONF(R)O(R)CH3(S)OH(R)HO

CAS RN 154361-50-9

IUPAC Name: pentyl N-{1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-methyloxolan-2-yl]-5-fluoro-2-oxo-1,2-dihydropyrimidin-4-yl}carbamate
Synonyms: capecitabine capecitabine (product) capecitabinum capecitabine (substance) capecitabina capecitabin capecitabine preparation 卡培他滨 xeloda CAPE
Molecular Weight (g/mol): 359.35
Molecular Formula: C15H22FN3O6
InChi Key: GAGWJHPBXLXJQN-UORFTKCHSA-N
SMILES: CCCCCOC(=O)NC1=NC(=O)N(C=C1F)[C@@H]1O[C@H](C)[C@@H](O)[C@H]1O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)