missing translation for 'onlineSavingsMsg'
Learn More

Capecitabine 98.0+%, TCI America™

Catalog No. C28785G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C28785G 5 g
C28781G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C28785G Supplier TCI America Supplier No. C28785G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 154361-50-9
Molecular Formula C15H22FN3O6
Molecular Weight (g/mol) 359.354
MDL Number MFCD00930626
InChI Key GAGWJHPBXLXJQN-UORFTKCHSA-N
Synonym capecitabine, xeloda, capiibine, capecitibine, caxeta, xabine, capecitabine usan, capecitabina, capecitabinum, capecitabin
PubChem CID 60953
ChEBI CHEBI:31348
IUPAC Name pentyl N-[1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-methyloxolan-2-yl]-5-fluoro-2-oxopyrimidin-4-yl]carbamate
SMILES CCCCCOC(=O)NC1=NC(=O)N(C=C1F)C2C(C(C(O2)C)O)O

Specifications

Melting Point 123°C
Color White
Quantity 5 g
Formula Weight 359.35
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Capecitabine
RTECSNumber HA3852500
TSCA No
Recommended Storage Refrigerator