missing translation for 'onlineSavingsMsg'
Learn More

Capecitabine Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013233
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013233 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013233 Supplier Merck Emd Millipore Supplier No. PHR14051G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Capecitabine is an orally administered drug belonging to the class of fluoropyrimidine carbamates. It converts into 5-fluorouracil (5-FU) at the tumor tissues by exploiting the high intratumoral concentrations of thymidine phosphorylase. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 154361-50-9
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C15H22 FN3O6
MDL Number MFCD00930626
Synonym Capecitabine; 5′-Deoxy-5-fluoro-N-[(pentyloxy)carbonyl]cytidine; Ro-9-1978
Type Certified Reference Material
Quantity 1 g
Formula Weight 359.35
Grade Certified Reference Material