accessibility menu, dialog, popup

UserName

CAS RN 154229-18-2

IUPAC Name: (7S,9aR,11aS)-9a,11a-dimethyl-1-(pyridin-3-yl)-3H,3aH,3bH,4H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-yl acetate
Synonyms: (7S,9aR,11aS)-9a,11a-dimethyl-1-(pyridin-3-yl)-3H,3aH,3bH,4H,6H,7H,8H,9H,9bH,10H,11H-cyclopenta[a]phenanthren-7-yl acetate
Molecular Weight (g/mol): 391.56
Molecular Formula: C26H33NO2
InChi Key: UVIQSJCZCSLXRZ-HMMZIKKISA-N
SMILES: CC(=O)O[C@H]1CC[C@]2(C)C3CC[C@@]4(C)C(CC=C4C4=CC=CN=C4)C3CC=C2C1
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results