CAS RN 154170-44-2
IUPAC Name:
4-methylnonan-5-ol
Synonyms:
4-methylnonan-5-ol
Molecular Weight (g/mol):
158.29
Molecular Formula:
C10H22O
InChi Key:
MBZNNOPVFZCHID-UHFFFAOYNA-N
SMILES:
CCCCC(O)C(C)CCC
1
–
1
of
1
results
1
Promotions Available
4-Methyl-5-nonanol, 98%
4-Methyl-5-nonanol, 98%
CAS: 154170-44-2 Molecular Formula: C10H22O Molecular Weight (g/mol): 158.29 MDL Number: MFCD00046696 InChI Key: MBZNNOPVFZCHID-UHFFFAOYNA-N Synonym: 4-methyl-5-nonanol,5-nonanol, 4-methyl,4-methyl 5-nonanol,acmc-1c1h3 PubChem CID: 3025865 IUPAC Name: 4-methylnonan-5-ol SMILES: CCCCC(O)C(C)CCC