accessibility menu, dialog, popup

UserName

H3COONNH2HNO

CAS RN 15400-53-0

IUPAC Name: ethyl 2-amino-6-oxo-1,6-dihydropyrimidine-5-carboxylate
Synonyms: ethyl 2-amino-4-oxo-3H-pyrimidine-5-carboxylate
Molecular Weight (g/mol): 183.17
Molecular Formula: C7H9N3O3
InChi Key: HRRHGLKNOJHIGY-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CN=C(N)NC1=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results