accessibility menu, dialog, popup

UserName

CAS RN 154-58-5

IUPAC Name: (2R,3S,4R,5S)-2-(hydroxymethyl)oxane-3,4,5-triol
Synonyms: 1,5-anhydroglucitol anhydroglucitol
Molecular Weight (g/mol): 164.16
Molecular Formula: C6H12O5
InChi Key: MPCAJMNYNOGXPB-SLPGGIOYSA-N
SMILES: OC[C@H]1OC[C@H](O)[C@@H](O)[C@@H]1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results