accessibility menu, dialog, popup

UserName

CAS RN 1538604-68-0

CH3NHOH2CNHNNClOCH3OCH3Cl

CAS RN 1538604-68-0

IUPAC Name: N-(2-{[6-(2,6-dichloro-3,5-dimethoxyphenyl)quinazolin-2-yl]amino}-3-methylphenyl)prop-2-enamide
Synonyms: N-(2-{[6-(2,6-dichloro-3,5-dimethoxyphenyl)quinazolin-2-yl]amino}-3-methylphenyl)prop-2-enamide
Molecular Weight (g/mol): 509.39
Molecular Formula: C26H22Cl2N4O3
InChi Key: TXEBNKKOLVBTFK-UHFFFAOYSA-N
SMILES: COC1=CC(OC)=C(Cl)C(=C1Cl)C1=CC2=CN=C(NC3=C(C)C=CC=C3NC(=O)C=C)N=C2C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results