missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BLU9931, 95%

Catalog No. AC468111000
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC468111000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC468111000 Supplier Thermo Scientific™ Supplier No. 468111000

Specifications

Chemical Name or Material BLU9931
CAS 1538604-68-0
Synonym N-(2-((6-(2,6-Dichloro-3,5-dimethoxyphenyl)quinazolin-2-yl)amino)-3-methylphenyl)acrylamide
Quantity 100 mg
Molecular Formula C26 H22 Cl2 N4 O3
Formula Weight 509.39
Physical Form Solid
Product Type Reagent
Purity 95%