CAS RN 1536327-95-3
CAS RN 1536327-95-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
2
Azido-PEG3-carboxyrhodamine 110 conjugate, Thermo Scientific Chemicals
CAS: 1536327-95-3 Molecular Formula: C29H30N6O7 MDL Number: MFCD22380746 Synonym: Azide-Fluor 488