accessibility menu, dialog, popup

UserName

CAS RN 153624-46-5

IUPAC Name: [4-(propan-2-yloxy)phenyl]boronic acid
Synonyms: 4-isopropoxyphenylboronic acid
Molecular Weight (g/mol): 180.01
Molecular Formula: C9H13BO3
InChi Key: CJUHQADBFQRIMC-UHFFFAOYSA-N
SMILES: CC(C)OC1=CC=C(C=C1)B(O)O
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)