CAS RN 15310-28-8
IUPAC Name:
1-methyl-4-[(4-methylbenzenesulfonyl)methanesulfonyl]benzene
Synonyms:
1-methyl-4-[(4-methylbenzenesulfonyl)methanesulfonyl]benzene
Molecular Weight (g/mol):
324.41
Molecular Formula:
C15H16O4S2
InChi Key:
XLOXYWLRAKCDQL-UHFFFAOYSA-N
SMILES:
CC1=CC=C(C=C1)S(=O)(=O)CS(=O)(=O)C1=CC=C(C)C=C1
1
–
1
of
1
results
1
Promotions Available
Bis(p-toluenesulfonyl)methane, 97%
Bis(p-toluenesulfonyl)methane, 97%
CAS: 15310-28-8 Molecular Formula: C15H16O4S2 Molecular Weight (g/mol): 324.41 MDL Number: MFCD00032255 InChI Key: XLOXYWLRAKCDQL-UHFFFAOYSA-N Synonym: ditosylmethane,bis-toluene-4-sulfonyl-methane,1-methyl-4-4-methylbenzenesulfonyl methanesulfonyl benzene,1,1'-methanediyldisulfonyl bis 4-methylbenzene,acmc-20allu,bis p-tolylsulfonyl methane,bis p-toluenesulfonyl methane,bis-toluene-4-sulfonyl methane,1,1'-methylenedisulfonyl bis 4-methylbenzene,benzene,1,1'-methylenebis sulfonyl bis 4-methyl PubChem CID: 275690 IUPAC Name: 1-methyl-4-[(4-methylphenyl)sulfonylmethylsulfonyl]benzene SMILES: CC1=CC=C(C=C1)S(=O)(=O)CS(=O)(=O)C1=CC=C(C)C=C1