accessibility menu, dialog, popup

UserName

CAS RN 153034-94-7

IUPAC Name: 2-fluoro-4-iodo-5-methylpyridine
Synonyms: 2-fluoro-4-iodo-5-methylpyridine
Molecular Weight (g/mol): 237.02
Molecular Formula: C6H5FIN
InChi Key: BVKQLNXPPQEELX-UHFFFAOYSA-N
SMILES: CC1=CN=C(F)C=C1I
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results