accessibility menu, dialog, popup

UserName

CAS RN 15301-40-3

IUPAC Name: 8-ethoxyquinoline-5-sulfonic acid
Synonyms: actinoquinol 阿克汀喹
Molecular Weight (g/mol): 253.27
Molecular Formula: C11H11NO4S
InChi Key: YAMVZYRZAMBCED-UHFFFAOYSA-N
SMILES: CCOC1=C2N=CC=CC2=C(C=C1)S(O)(=O)=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)