accessibility menu, dialog, popup

UserName

BrCH3OOP

CAS RN 1530-45-6

IUPAC Name: (2-ethoxy-2-oxoethyl)triphenylphosphanium bromide
Synonyms: (2-ethoxy-2-oxoethyl)triphenylphosphanium bromide
Molecular Weight (g/mol): 429.29
Molecular Formula: C22H22BrO2P
InChi Key: VJVZPTPOYCJFNI-UHFFFAOYSA-M
SMILES: [Br-].CCOC(=O)C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)