missing translation for 'onlineSavingsMsg'
Learn More

Ethoxycarbonylmethyl(triphenyl)phosphonium Bromide 97.0+%, TCI America™

Catalog No. E040725G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E040725G 25 g
E0407250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E040725G Supplier TCI America Supplier No. E040725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1530-45-6
Molecular Formula C22H22BrO2P
Molecular Weight (g/mol) 429.29
MDL Number MFCD00011835
InChI Key VJVZPTPOYCJFNI-UHFFFAOYSA-M
Synonym carbethoxymethyl triphenylphosphonium bromide, 2-ethoxy-2-oxoethyl triphenylphosphonium bromide, ethoxycarbonylmethyl triphenylphosphonium bromide, carbethoxymethylenetriphenylphosphonium bromide, ethoxycarbonylmethyl triphenyl phosphonium bromide, phosphonium, 2-ethoxy-2-oxoethyl triphenyl-, bromide, carbethoxymethyl-triphenyl-phosphonium bromide, 2-ethoxy-2-oxoethyl triphenylphosphanium bromide, carboethoxymethyl triphenylphosphonium bromide, phosphonium, ethoxycarbonylmethyl triphenyl-, bromide
PubChem CID 73731
IUPAC Name (2-ethoxy-2-oxoethyl)triphenylphosphanium bromide
SMILES [Br-].CCOC(=O)C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 158°C
Quantity 25 g
Formula Weight 429.29
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Ethoxycarbonylmethyl(triphenyl)phosphonium Bromide
RTECSNumber TA2299500
TSCA No