missing translation for 'onlineSavingsMsg'
Learn More

(Ethoxycarbonylmethyl)triphenylphosphonium bromide, 98+%

Catalog No. AAA1634736
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1634736 500 g
AAA1634714 25 g
AAA1634722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1634736 Supplier Thermo Scientific Chemicals Supplier No. A1634736
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1530-45-6 | C22H22BrO2P | 429.29 g/mol

(Ethoxycarbonylmethyl)triphenylphosphonium bromide, is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(Ethoxycarbonylmethyl)triphenylphosphonium bromide, is used as a pharmaceutical intermediate.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents, Strong bases.

Chemical Identifiers

CAS 1530-45-6
Molecular Formula C22H22BrO2P
Molecular Weight (g/mol) 429.29
MDL Number MFCD00011835
InChI Key VJVZPTPOYCJFNI-UHFFFAOYSA-M
Synonym carbethoxymethyl triphenylphosphonium bromide, 2-ethoxy-2-oxoethyl triphenylphosphonium bromide, ethoxycarbonylmethyl triphenylphosphonium bromide, carbethoxymethylenetriphenylphosphonium bromide, ethoxycarbonylmethyl triphenyl phosphonium bromide, phosphonium, 2-ethoxy-2-oxoethyl triphenyl-, bromide, carbethoxymethyl-triphenyl-phosphonium bromide, 2-ethoxy-2-oxoethyl triphenylphosphanium bromide, carboethoxymethyl triphenylphosphonium bromide, phosphonium, ethoxycarbonylmethyl triphenyl-, bromide
PubChem CID 73731
SMILES [Br-].CCOC(=O)C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point ∼150°C (decomposition)
Quantity 500 g
Beilstein 3581273
Sensitivity Hygroscopic
Solubility Information It is soluble in water.
IUPAC Name (2-ethoxy-2-oxoethyl)-triphenylphosphanium;bromide
Formula Weight 429.3
Percent Purity ≥98%
Chemical Name or Material (Ethoxycarbonylmethyl)triphenylphosphonium bromide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 216-230-1
RTECSNumber TA2299500
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only