CAS RN 15291-75-5
IUPAC Name:
8-tert-butyl-6,17-dihydroxy-16-methyl-2,4,14,19-tetraoxahexacyclo[8.7.2.0¹,¹¹.0³,⁷.0⁷,¹¹.0¹³,¹⁷]nonadecane-5,15,18-trione
Synonyms:
8-tert-butyl-6,17-dihydroxy-16-methyl-2,4,14,19-tetraoxahexacyclo[8.7.2.0¹,¹¹.0³,⁷.0⁷,¹¹.0¹³,¹⁷]nonadecane-5,15,18-trione
Molecular Weight (g/mol):
408.4
Molecular Formula:
C20H24O9
InChi Key:
FPUXKXIZEIDQKW-UHFFFAOYNA-N
SMILES:
CC1C(=O)OC2CC34C5CC(C(C)(C)C)C33C(O)C(=O)OC3OC4(C(=O)O5)C12O
1
–
3
of
3
results
2
Ginkgolide A from Ginkgo biloba leaves, 90% (HPLC), MilliporeSigma™ Supelco™
Ginkgolide A is a diterpene lactone, which can be extracted from the leaves of Ginkgo biloba tree.
3
Ginkgolide A, Thermo Scientific Chemicals
CAS: 15291-75-5 Molecular Formula: C20H24O9 Molecular Weight (g/mol): 408.40 MDL Number: MFCD00133365,MFCD07437829 InChI Key: FPUXKXIZEIDQKW-UHFFFAOYNA-N Synonym: ginkgolide a PubChem CID: 132470876 IUPAC Name: 8-tert-butyl-6,17-dihydroxy-16-methyl-2,4,14,19-tetraoxahexacyclo[8.7.2.0¹,¹¹.0³,⁷.0⁷,¹¹.0¹³,¹⁷]nonadecane-5,15,18-trione SMILES: CC1C(=O)OC2CC34C5CC(C(C)(C)C)C33C(O)C(=O)OC3OC4(C(=O)O5)C12O