missing translation for 'onlineSavingsMsg'
Learn More

Ginkgolide A, Thermo Scientific Chemicals

Catalog No. AAJ62384FPL
Change view
Click to view available options
Quantity:
20 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62384FPL 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ62384FPL Supplier Thermo Scientific Chemicals Supplier No. J62384FPL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

 

Platelet-Activating Factor (PAF) antagonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15291-75-5
Molecular Formula C20H24O9
Molecular Weight (g/mol) 408.40
MDL Number MFCD00133365,MFCD07437829
InChI Key FPUXKXIZEIDQKW-UHFFFAOYNA-N
Synonym ginkgolide a
PubChem CID 132470876
IUPAC Name 8-tert-butyl-6,17-dihydroxy-16-methyl-2,4,14,19-tetraoxahexacyclo[8.7.2.0¹,¹¹.0³,⁷.0⁷,¹¹.0¹³,¹⁷]nonadecane-5,15,18-trione
SMILES CC1C(=O)OC2CC34C5CC(C(C)(C)C)C33C(O)C(=O)OC3OC4(C(=O)O5)C12O

Specifications

Color White
Quantity 20 mg
Merck Index 14,4426
Solubility Information Soluble in DMSO
Formula Weight 408.41
Physical Form Powder
Chemical Name or Material Ginkgolide A
RTECSNumber KC9943000
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C; Store under Argon

RUO – Research Use Only