accessibility menu, dialog, popup

UserName

H3CONH2NON

CAS RN 1527503-11-2

IUPAC Name: 5'-methoxy-6'-[3-(pyrrolidin-1-yl)propoxy]spiro[cyclobutane-1,3'-indol]-2'-amine
Synonyms: 5'-methoxy-6'-[3-(pyrrolidin-1-yl)propoxy]spiro[cyclobutane-1,3'-indol]-2'-amine
Molecular Weight (g/mol): 329.44
Molecular Formula: C19H27N3O2
InChi Key: BKCDJTRMYWSXMC-UHFFFAOYSA-N
SMILES: COC1=C(OCCCN2CCCC2)C=C2N=C(N)C3(CCC3)C2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results