missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ A-366, 95%

Catalog No. AC468071000
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC468071000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC468071000 Supplier Thermo Scientific™ Supplier No. 468071000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material A-366
CAS 1527503-11-2
Synonym 5 '-Methoxy-6 '-[3-(1-pyrrolidinyl)propoxy]-spiro[cyclobutane-1,3 '-[3 H]indol]-2 '-amine
Quantity 100 mg
Molecular Formula C19 H27 N3 O2
Formula Weight 329.44
Physical Form Solid
Product Type Reagent
Purity 95%