accessibility menu, dialog, popup

UserName

CAS RN 15263-20-4

H3CCH3H3COOONOO

CAS RN 15263-20-4

IUPAC Name: tert-butyl 1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl carbonate
Synonyms: tert-butyl 1,3-dioxoisoindol-2-yl carbonate
Molecular Weight (g/mol): 263.25
Molecular Formula: C13H13NO5
InChi Key: MCWXBNWFVFOQAS-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)ON1C(=O)C2=CC=CC=C2C1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results