accessibility menu, dialog, popup

UserName

CH3NOONNH2NH2NOOHO

CAS RN 152460-08-7

IUPAC Name: N''-(2-methyl-5-nitrophenyl)guanidine; nitric acid
Synonyms: N''-(2-methyl-5-nitrophenyl)guanidine; nitric acid
Molecular Weight (g/mol): 257.21
Molecular Formula: C8H11N5O5
InChi Key: IINMQQJNRFDBMV-UHFFFAOYSA-N
SMILES: O[N+]([O-])=O.CC1=C(C=C(C=C1)[N+]([O-])=O)N=C(N)N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results