CAS RN 152459-95-5
IUPAC Name:
N-(4-methyl-3-{[4-(pyridin-3-yl)pyrimidin-2-yl]amino}phenyl)-4-[(4-methylpiperazin-1-yl)methyl]benzamide
Synonyms:
imatinib imatinib preparation imatinib (substance) imatinib mesylate preparation genpharm brand of imatinib mesylate imatinib methansulfonate imatinib (product) imatinib mesylate imatinib mesylate (substance) novartis brand of imatinib mesylate
Molecular Weight (g/mol):
493.62
Molecular Formula:
C29H31N7O
InChi Key:
KTUFNOKKBVMGRW-UHFFFAOYSA-N
SMILES:
CN1CCN(CC2=CC=C(C=C2)C(=O)NC2=CC=C(C)C(NC3=NC=CC(=N3)C3=CC=CN=C3)=C2)CC1
1
–
1
of
1
results
1
Imatinib 98.0+%, TCI America™
CAS: 152459-95-5 Molecular Formula: C29H31N7O Molecular Weight (g/mol): 493.62 MDL Number: MFCD05662257 InChI Key: KTUFNOKKBVMGRW-UHFFFAOYSA-N PubChem CID: 5291 ChEBI: CHEBI:45783 IUPAC Name: N-(4-methyl-3-{[4-(pyridin-3-yl)pyrimidin-2-yl]amino}phenyl)-4-[(4-methylpiperazin-1-yl)methyl]benzamide SMILES: CN1CCN(CC2=CC=C(C=C2)C(=O)NC2=CC=C(C)C(NC3=NC=CC(=N3)C3=CC=CN=C3)=C2)CC1