accessibility menu, dialog, popup

UserName

CH3HOFFFFFF

CAS RN 1515-14-6

IUPAC Name: 1,1,1,3,3,3-hexafluoro-2-methylpropan-2-ol
Synonyms: 1,1,1,3,3,3-hexafluoro-2-methylpropan-2-ol
Molecular Weight (g/mol): 182.07
Molecular Formula: C4H4F6O
InChi Key: FQDXJYBXPOMIBX-UHFFFAOYSA-N
SMILES: CC(O)(C(F)(F)F)C(F)(F)F
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)