missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,3,3,3-Hexafluoro-2-methyl-2-propanol, 98%

Catalog No. AAL0924514
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0924514 25 g
AAL0924506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0924514 Supplier Thermo Scientific Chemicals Supplier No. L0924514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1515-14-6 | C4H4F6O | 182.07 g/mol

1,1,1,3,3,3-Hexafluoro-2-methyl-2-propanol is used as a solvent. It is used in the preparation of volatile sodium yttrium and zirconium fluoroalkoxides. It is also used in the synthesis of precursors to per- and polyfluoroethers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1,1,3,3,3-Hexafluoro-2-methyl-2-propanol is used as a solvent. It is used in the preparation of volatile sodium yttrium and zirconium fluoroalkoxides. It is also used in the synthesis of precursors to per- and polyfluoroethers.

Solubility
Slightly soluble in water.

Notes
Incompatible materials are strong acids, strong bases, strong oxidizing agents, strong reducing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 1515-14-6
Molecular Formula C4H4F6O
Molecular Weight (g/mol) 182.07
MDL Number MFCD00039259
InChI Key FQDXJYBXPOMIBX-UHFFFAOYSA-N
Synonym hexafluoro-2-methylisopropanol, 1,1,1,3,3,3-hexafluoro-2-methyl-2-propanol, 2-propanol, 1,1,1,3,3,3-hexafluoro-2-methyl, hexafluoro-tert-butanol, cf3 2mecoh, 1,1,1,3,3,3-hexafluoro-2-methylisopropanol, methylbistrifluoromethylmethanol, cf3 2c oh ch3, fqdxjybxpomibx-uhfffaoysa, hexafluoro-3-methylisopropanol
PubChem CID 73936
IUPAC Name 1,1,1,3,3,3-hexafluoro-2-methylpropan-2-ol
SMILES CC(O)(C(F)(F)F)C(F)(F)F

Specifications

Density 1.484
Boiling Point 60°C to 62°C
Refractive Index 1.295
Quantity 25 g
Beilstein 1841110
Solubility Information Slightly soluble in water.
Formula Weight 182.07
Percent Purity 98%
Chemical Name or Material 1,1,1,3,3,3-Hexafluoro-2-methyl-2-propanol
Hazard Category H302-H312-H315-H319-H332-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 216-155-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only