missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1,1,3,3,3-Hexafluoro-2-méthyl-2-propanol 98,0+%, TCI America™
Spécifications
Spécifications
| Color | Incolore |
| Boiling Point | 62°C |
| Quantity | 5 g |
| Formula Weight | 182.07 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquide |
| Chemical Name or Material | 1,1,1,3,3,3-Hexafluoro-2-methyl-2-propanol |
Sécurité et manipulation
| TSCA | Yes |