accessibility menu, dialog, popup

UserName

CAS RN 151486-56-5

BNNH3CH3CH3CCH3CH3CH3CH3FF

CAS RN 151486-56-5

IUPAC Name: 5,11-dibutyl-2,2-difluoro-4,6,8,10,12-pentamethyl-1λ⁵,3-diaza-2-boratricyclo[7.3.0.0³,⁷]dodeca-1(12),4,6,8,10-pentaen-1-ylium-2-uide
Synonyms: 5,11-dibutyl-2,2-difluoro-4,6,8,10,12-pentamethyl-1λ⁵,3-diaza-2-boratricyclo[7.3.0.0³,⁷]dodeca-1(12),4,6,8,10-pentaen-1-ylium-2-uide
Molecular Weight (g/mol): 374.33
Molecular Formula: C22H33BF2N2
InChi Key: XWUUMDJCWNZHAZ-UHFFFAOYSA-N
SMILES: CCCCC1=C(C)N2C(=C1C)C(C)=C1C(C)=C(CCCC)C(C)=[N+]1[B-]2(F)F
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results