accessibility menu, dialog, popup

UserName

CAS RN 15104-46-8

IUPAC Name: dimethyl [2-(dimethoxyphosphoryl)phenyl]phosphonate
Synonyms: dimethyl 2-(dimethoxyphosphoryl)phenylphosphonate
Molecular Weight (g/mol): 294.18
Molecular Formula: C10H16O6P2
InChi Key: TUKTVDDATWNXSN-UHFFFAOYSA-N
SMILES: COP(=O)(OC)C1=CC=CC=C1P(=O)(OC)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results