accessibility menu, dialog, popup

UserName

CAS RN 15055-81-9

IUPAC Name: methyl 1,2-oxazole-5-carboxylate
Synonyms: methyl 1,2-oxazole-5-carboxylate
Molecular Weight (g/mol): 127.1
Molecular Formula: C5H5NO3
InChi Key: ILPCPKZAAQXHKL-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=NO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results