accessibility menu, dialog, popup

UserName

CH3H3COOOHOOOH

CAS RN 15042-01-0

IUPAC Name: 2-(2,2-dimethyl-1,3-dioxolan-4-yl)-4,5-dihydroxy-2,3-dihydrofuran-3-one
Synonyms: 2-(2,2-dimethyl-1,3-dioxolan-4-yl)-4,5-dihydroxy-2H-furan-3-one
Molecular Weight (g/mol): 216.19
Molecular Formula: C9H12O6
InChi Key: KLQRPFQLSOJOQI-UHFFFAOYNA-N
SMILES: CC1(C)OCC(O1)C1OC(O)=C(O)C1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results