accessibility menu, dialog, popup

UserName

CAS RN 1504-63-8

IUPAC Name: (2E)-3-(4-nitrophenyl)prop-2-en-1-ol
Synonyms: (2E)-3-(4-nitrophenyl)prop-2-en-1-ol
Molecular Weight (g/mol): 179.18
Molecular Formula: C9H9NO3
InChi Key: LGXXEDSIJZHDBN-OWOJBTEDSA-N
SMILES: OC\C=C\C1=CC=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results