accessibility menu, dialog, popup

UserName

O

CAS RN 1502-22-3

IUPAC Name: 2-(cyclohex-1-en-1-yl)cyclohexan-1-one
Synonyms: 2-cyclohexenyl cyclohexanone
Molecular Weight (g/mol): 178.28
Molecular Formula: C12H18O
InChi Key: GVNVAWHJIKLAGL-UHFFFAOYNA-N
SMILES: O=C1CCCCC1C1=CCCCC1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)