accessibility menu, dialog, popup

UserName

CAS RN 14995-22-3

IUPAC Name: iron(3+) tris(propan-2-olate)
Synonyms: iron(3+) tris(propan-2-olate)
Molecular Weight (g/mol): 233.11
Molecular Formula: C9H21FeO3
InChi Key: QUHDSMAREWXWFM-UHFFFAOYSA-N
SMILES: [Fe+3].CC(C)[O-].CC(C)[O-].CC(C)[O-]
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)