accessibility menu, dialog, popup

UserName

HClHNN(E)(E)(E/Z)

CAS RN 1497-49-0

IUPAC Name: hydrogen N-[(1E,3E)-5-(phenylimino)penta-1,3-dien-1-yl]aniline chloride
Synonyms: hydrogen N-[(1E,3E)-5-(phenylimino)penta-1,3-dien-1-yl]aniline chloride
Molecular Weight (g/mol): 284.79
Molecular Formula: C17H17ClN2
InChi Key: VUCMMJBDNXZQDJ-ZUJIUJENSA-N
SMILES: [H+].[Cl-].N(\C=C\C=C\C=NC1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)