missing translation for 'onlineSavingsMsg'
Learn More

N-Hydroxy-N'-phenyloctanediamide 98.0+%, TCI America™

Catalog No. H1388200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H1388200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H1388200MG Supplier TCI America Supplier No. H1388200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 149647-78-9
Molecular Formula C14H20N2O3
Molecular Weight (g/mol) 264.325
MDL Number MFCD00945317
InChI Key WAEXFXRVDQXREF-UHFFFAOYSA-N
Synonym vorinostat, saha, suberoylanilide hydroxamic acid, zolinza, n-hydroxy-n'-phenyloctanediamide, suberanilohydroxamic acid, n1-hydroxy-n8-phenyloctanediamide, saha cpd, vorinostat usan, octanediamide, n-hydroxy-n'-phenyl
PubChem CID 5311
ChEBI CHEBI:45716
IUPAC Name N'-hydroxy-N-phenyloctanediamide
SMILES C1=CC=C(C=C1)NC(=O)CCCCCCC(=O)NO

Specifications

Melting Point 155°C
Color White
Quantity 200 mg
Formula Weight 264.33
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-Hydroxy-N′-phenyloctanediamide
RTECSNumber RG8835000
TSCA No
Recommended Storage Freezer (-20 °C)