accessibility menu, dialog, popup

UserName

CAS RN 14913-20-3

IUPAC Name: europium(3+) phosphate
Synonyms: europium(3+) phosphate
Molecular Weight (g/mol): 246.93
Molecular Formula: EuO4P
InChi Key: KJNZTHUWRRLWOA-UHFFFAOYSA-K
SMILES: [Eu+3].[O-]P([O-])([O-])=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results