accessibility menu, dialog, popup

UserName

CAS RN 14898-79-4

IUPAC Name: (2R)-butan-2-ol
Synonyms: (-)-2-butanol
Molecular Weight (g/mol): 74.12
Molecular Formula: C4H10O
InChi Key: BTANRVKWQNVYAZ-SCSAIBSYSA-N
SMILES: CC[C@@H](C)O
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)