accessibility menu, dialog, popup

UserName

HClNH2NH2NNH2OHO

CAS RN 1483-01-8

IUPAC Name: hydrogen 2-amino-6-[(diaminomethylidene)amino]hexanoic acid chloride
Synonyms: hydrogen homoarginine chloride
Molecular Weight (g/mol): 224.69
Molecular Formula: C7H17ClN4O2
InChi Key: YMKBVNVCKUYUDM-UHFFFAOYNA-N
SMILES: [H+].[Cl-].NC(CCCCN=C(N)N)C(O)=O
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (3)
  • (1)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (5)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)