accessibility menu, dialog, popup

UserName

CAS RN 1479-58-9

IUPAC Name: 4-bromo-2-(2-fluorobenzoyl)aniline
Synonyms: 4-bromo-2-(2-fluorobenzoyl)aniline
Molecular Weight (g/mol): 294.12
Molecular Formula: C13H9BrFNO
InChi Key: XCOKDXNGCQXFCV-UHFFFAOYSA-N
SMILES: NC1=CC=C(Br)C=C1C(=O)C1=CC=CC=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results