accessibility menu, dialog, popup

UserName

CAS RN 1479-24-9

H3COOOF

CAS RN 1479-24-9

IUPAC Name: ethyl 3-(2-fluorophenyl)-3-oxopropanoate
Synonyms: ethyl 3-(2-fluorophenyl)-3-oxopropanoate
Molecular Weight (g/mol): 210.2
Molecular Formula: C11H11FO3
InChi Key: YMUNUVJSNWUWDA-UHFFFAOYSA-N
SMILES: CCOC(=O)CC(=O)C1=CC=CC=C1F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results