accessibility menu, dialog, popup

UserName

CAS RN 14782-75-3

IUPAC Name: aluminium(3+) 1-ethoxy-1,3-dioxobutan-2-ide bis(propan-2-olate)
Synonyms: aluminium(3+) 1-ethoxy-1,3-dioxobutan-2-ide bis(propan-2-olate)
Molecular Weight (g/mol): 274.29
Molecular Formula: C12H23AlO5
InChi Key: AHPLRRISNPYWTH-UHFFFAOYSA-N
SMILES: [Al+3].CC(C)[O-].CC(C)[O-].CCOC(=O)[CH-]C(C)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)