accessibility menu, dialog, popup

UserName

CuFFFOHCOFFFFFFOHCOFFF

CAS RN 14781-45-4

IUPAC Name: copper(2+) bis(1,1,1,5,5,5-hexafluoro-2,4-dioxopentan-3-ide)
Synonyms: copper(2+) bis(1,1,1,5,5,5-hexafluoro-2,4-dioxopentan-3-ide)
Molecular Weight (g/mol): 477.65
Molecular Formula: C10H2CuF12O4
InChi Key: NPOQJGKWJAKNAV-UHFFFAOYSA-N
SMILES: [Cu++].FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F.FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results